CH6371379718 Chem Impex International, Inc.

Nd-Phthaloyl-L-ornithine hydrochloride

  • MDL number MFCD00190989
  • Cas Number : 10009-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-222
Synonym L-Orn(Pht)-OH HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-d-sup-phthaloyl-l-ornithine-hydrochloride/4362?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-d-sup-phthaloyl-l-ornithine-hydrochloride/4362?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart