CH6371379719 Chem Impex International, Inc.

Methoxycarbonyl-glycine

  • MDL number MFCD00270561
  • Cas Number : 1670-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-98
Synonym Methoxycarbonyl-Gly-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/methoxycarbonyl-glycine/4363?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/methoxycarbonyl-glycine/4363?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart