CH6371379720 Chem Impex International, Inc.

Benzyl-N-methyl-L-alanine

  • MDL number MFCD01632093
  • Cas Number : 63238-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Bzl-N-Me-L-Ala-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzyl-i-n-i-methyl-l-alanine/4366?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzyl-i-n-i-methyl-l-alanine/4366?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart