CH6371379724 Chem Impex International, Inc.

Fmoc-Gly-Pro-OH

  • MDL number MFCD00190883
  • Cas Number : 212651-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-gycyl-L-proline
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-gly-pro-oh/4371?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-gly-pro-oh/4371?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart