CH6371379725 Chem Impex International, Inc.

Tyr-Gly-OH

  • MDL number MFCD00037779
  • Cas Number : 673-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Tyrosylglycine
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/tyr-gly-oh/4373?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/tyr-gly-oh/4373?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart