CH6371379726 Chem Impex International, Inc.

Z-L-pyroglutamic acid dicyclohexylammonium salt

  • MDL number MFCD03788049
  • Cas Number : 38596-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-204
Synonym Z-L-Pyr-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-pyroglutamic-acid-dicyclohexylammonium-salt/4374?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-pyroglutamic-acid-dicyclohexylammonium-salt/4374?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart