CH6371379730 Chem Impex International, Inc.

Boc-O-methyl-L-tyrosine methyl ester

  • MDL number MFCD03788050
  • Cas Number : 94790-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Tyr(Me)-OMe
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-methyl-l-tyrosine-methyl-ester/4382?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-methyl-l-tyrosine-methyl-ester/4382?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart