CH6371379733 Chem Impex International, Inc.

Acetoacetyl coenzyme A lithium salt

  • MDL number MFCD00038868

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at = -15
Physical Form Solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetoacetyl-coenzyme-a-lithium-salt/4398?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetoacetyl-coenzyme-a-lithium-salt/4398?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart