CH6371379739 Chem Impex International, Inc.

Fmoc-S-acetamidomethyl-L-homocysteine

  • MDL number MFCD03788052
  • Cas Number : 150281-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-HomoCys(Acm)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-s-acetamidomethyl-l-homocysteine/4405?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-s-acetamidomethyl-l-homocysteine/4405?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart