CH6371379745 Chem Impex International, Inc.

S-4-Methoxybenzyl-L-penicillamine

  • MDL number MFCD03428497
  • Cas Number : 387868-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-171
Synonym L-Pen(pMeOBzl)-OH
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-4-methoxybenzyl-l-penicillamine/4416?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-4-methoxybenzyl-l-penicillamine/4416?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart