CH6371379747 Chem Impex International, Inc.

S-4-Methylbenzyl-D-penicillamine

  • MDL number MFCD03788057
  • Cas Number : 1330286-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168-172
Synonym D-Pen(pMeBzl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-4-methylbenzyl-d-penicillamine/4418?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-4-methylbenzyl-d-penicillamine/4418?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart