CH6371379762 Chem Impex International, Inc.

Fmoc-O-methyl-D-tyrosine

  • MDL number MFCD00237393
  • Cas Number : 201335-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-166
Synonym Fmoc-D-Tyr(Me)-OH
Purity 99.7
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-methyl-d-tyrosine/4434?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-methyl-d-tyrosine/4434?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart