CH6371379764 Chem Impex International, Inc.

Na-Fmoc-Ne-(Boc-2-aminobenzoyl)-L-lysine

  • MDL number MFCD01318747
  • Cas Number : 159322-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-ablysin(Boc)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-boc-2-aminobenzoyl-l-lysine/4438?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-boc-2-aminobenzoyl-l-lysine/4438?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart