CH6371379780 Chem Impex International, Inc.

Ac-4,5-dehydro-L-leucine

  • MDL number MFCD00237465
  • Cas Number : 88547-24-4
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-175 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White product
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ac-4-5-dehydro-l-leucine/4462?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ac-4-5-dehydro-l-leucine/4462?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart