CH6371379781 Chem Impex International, Inc.

Nalpha-Acetyl-5-methyl-DL-tryptophan

  • MDL number MFCD00055984
  • Cas Number : 71953-90-7
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-170 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Off-white product
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-acetyl-5-methyl-dl-tryptophan/4465?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-acetyl-5-methyl-dl-tryptophan/4465?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart