CH6371379783 Chem Impex International, Inc.

Fmoc-D-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid

  • MDL number MFCD03788061
  • Cas Number : 204317-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form Amorphous white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-1-2-3-4-tetrahydroisoquinoline-1-carboxylic-acid/4467?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-1-2-3-4-tetrahydroisoquinoline-1-carboxylic-acid/4467?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart