CH6371379786 Chem Impex International, Inc.

Acetyl-DL-propargylglycine ethyl ester

  • MDL number MFCD00237477
  • Cas Number : 23235-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-DL-propargyl-Gly-OEt
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-dl-propargylglycine-ethyl-ester/4470?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-dl-propargylglycine-ethyl-ester/4470?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart