CH6371379789 Chem Impex International, Inc.

5-Aminovaleric acid benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00237402
  • Cas Number : 63649-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63 - 69
Synonym 5-Ava-OBzl TosOH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-aminovaleric-acid-benzyl-ester-4-toluenesulfonate-salt/4473?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-aminovaleric-acid-benzyl-ester-4-toluenesulfonate-salt/4473?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart