CH6371379791 Chem Impex International, Inc.

Boc-4-tert-butyl-L-phenylalanine dicyclohexylammonium salt

  • MDL number MFCD19707042

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Phe(4-tBu)-OH DCHA
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-i-tert-i-butyl-l-phenylalanine-dicyclohexylammonium-salt/4475?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-i-tert-i-butyl-l-phenylalanine-dicyclohexylammonium-salt/4475?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart