CH6371379794 Chem Impex International, Inc.

Nalpha-Boc-L-2,4-diaminobutyric acid tert-butyl ester hydrochloride

  • MDL number MFCD02259473
  • Cas Number : 190447-69-9
  • Molecular Weight : 310.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-l-2-4-diaminobutyric-acid-i-tert-i-butyl-ester-hydrochloride/4478?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-l-2-4-diaminobutyric-acid-i-tert-i-butyl-ester-hydrochloride/4478?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart