CH6371379803 Chem Impex International, Inc.

4-Carboxy-L-phenylalanine

  • MDL number MFCD01318768
  • Cas Number : 126109-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Phe(4-COOH)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-carboxy-l-phenylalanine/4492?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-carboxy-l-phenylalanine/4492?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart