CH6371379811 Chem Impex International, Inc.

b-Fluoro-DL-alanine

  • MDL number MFCD00077524
  • Cas Number : 43163-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-149
Synonym DL-beta-FluoroAla-OH
Purity 97
Storage temp. Store at 0-8
Physical Form Solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/beta-fluoro-dl-alanine/4503?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/beta-fluoro-dl-alanine/4503?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart