CH6371379812 Chem Impex International, Inc.

4-Fluoro-L-phenylalanine ethyl ester hydrochloride

  • MDL number MFCD02259530
  • Cas Number : 1534-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-153
Synonym L-Phe(4-F)-OEt HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-fluoro-l-phenylalanine-ethyl-ester-hydrochloride/4504?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-fluoro-l-phenylalanine-ethyl-ester-hydrochloride/4504?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart