CH6371379813 Chem Impex International, Inc.

Fmoc-L-allo-threonine

  • MDL number MFCD01318739
  • Cas Number : 146306-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-i-allo-i-threonine/4505?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-i-allo-i-threonine/4505?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart