CH6371379816 Chem Impex International, Inc.

Fmoc-4-tert-butyl-L-phenylalanine

  • MDL number MFCD02094466
  • Cas Number : 213383-02-9
  • Molecular Weight : 443.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-i-tert-i-butyl-l-phenylalanine/4508?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-i-tert-i-butyl-l-phenylalanine/4508?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart