CH6371379819 Chem Impex International, Inc.

Fmoc-S-Boc-3-aminopropyl-L-cysteine

  • MDL number MFCD01318742
  • Cas Number : 173963-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-119 �C
Synonym Fmoc-L-Cys(Boc-3-aminopropyl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-boc-3-aminopropyl-l-cysteine/4511?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-boc-3-aminopropyl-l-cysteine/4511?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart