CH6371379825 Chem Impex International, Inc.

Fmoc-DL-isoserine

  • MDL number MFCD00672327
  • Cas Number : 161125-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-160
Synonym Fmoc-DL-isoserine
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-dl-isoserine/4517?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-dl-isoserine/4517?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart