CH6371379830 Chem Impex International, Inc.

(2S,4S)-g-Hydroxy-L-glutamic acid

  • MDL number MFCD00672374
  • Cas Number : 3913-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2S,4S)-gamma-Hydroxy-L-Glu-OH
Purity 96
Storage temp. Store at 0-8
Physical Form White product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-i-s-i-4-i-s-i-gamma-hydroxy-l-glutamic-acid/4522?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-i-s-i-4-i-s-i-gamma-hydroxy-l-glutamic-acid/4522?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart