CH6371379834 Chem Impex International, Inc.

4-Methyl-L-phenylalanine

  • MDL number MFCD00038575
  • Cas Number : 1991-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Phe(4-Me)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methyl-l-phenylalanine/4526?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methyl-l-phenylalanine/4526?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart