CH6371379845 Chem Impex International, Inc.

3-(2'-Quinolyl)-D-alanine

  • MDL number MFCD01318775
  • Cas Number : 170421-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Ala(2'-quinolyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Gray powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-2-quinolyl-d-alanine/4539?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-2-quinolyl-d-alanine/4539?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart