CH6371379847 Chem Impex International, Inc.

O-Sulfo-L-serine

  • MDL number MFCD00672378
  • Cas Number : 626-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-214 °C
Purity 101
Storage temp. Store at 0-8 °C
Physical Form White crystals
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-sulfo-l-serine/4541?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-sulfo-l-serine/4541?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart