CH6371379849 Chem Impex International, Inc.

DL-Thyronine

  • MDL number MFCD00045864
  • Cas Number : 1034-10-2
  • Molecular Weight : 273.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-thyronine/4544?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-thyronine/4544?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart