CH6371379850 Chem Impex International, Inc.

(3R)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid tert-butyl ester hydrochloride

  • MDL number MFCD00672372
  • Cas Number : 103733-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228-232
Synonym D-Tic-OtBu HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-i-r-i-1-2-3-4-tetrahydroisoquinoline-3-carboxylic-acid-i-tert-i-butyl-ester-hydrochloride/4545?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-i-r-i-1-2-3-4-tetrahydroisoquinoline-3-carboxylic-acid-i-tert-i-butyl-ester-hydrochloride/4545?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart