CH6371379854 Chem Impex International, Inc.

Z-b-cyclohexyl-D-alanine dicyclohexylammonium salt

  • MDL number MFCD01862304
  • Cas Number : 214852-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-161
Synonym Z-D-Cha-OH DCHA
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-beta-cyclohexyl-d-alanine-dicyclohexylammonium-salt/4549?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-beta-cyclohexyl-d-alanine-dicyclohexylammonium-salt/4549?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart