CH6371379855 Chem Impex International, Inc.

Na-Z-Ng-Boc-D-2,4-diaminobutyric acid dicyclohexylammonium salt

  • MDL number MFCD01318750
  • Cas Number : 214852-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-196
Synonym Z-D-Dab(Boc)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-gamma-sup-boc-d-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/4550?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-gamma-sup-boc-d-2-4-diaminobutyric-acid-dicyclohexylammonium-salt/4550?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart