CH6371379865 Chem Impex International, Inc.

Fmoc-O-trityl-D-threonine

  • MDL number MFCD01631661
  • Cas Number : 682800-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-75
Synonym Fmoc-D-Thr(Trt)-OH
Purity 98
Storage temp. Store at = -4
Physical Form Off-white to brown crystals
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-trityl-d-threonine/4561?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-trityl-d-threonine/4561?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart