CH6371379871 Chem Impex International, Inc.

Boc-4-aminomethylcyclohexane carboxylic acid

  • MDL number MFCD01862289
  • Cas Number : 162046-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-137 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-aminomethylcyclohexane-carboxylic-acid/4567?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-aminomethylcyclohexane-carboxylic-acid/4567?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart