CH6371379883 Chem Impex International, Inc.

Acetyl-L-norleucine

  • MDL number MFCD00151961
  • Cas Number : 15891-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-110
Synonym Ac-L-Nle-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-norleucine/4579?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-norleucine/4579?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart