CH6371379884 Chem Impex International, Inc.

Boc-4-(Z-amino)-D-phenylalanine dicyclohexylammonium salt

  • MDL number MFCD02092967

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Phe(4-NHZ)-OH DCHA
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-z-amino-d-phenylalanine-dicyclohexylammonium-salt/4580?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-z-amino-d-phenylalanine-dicyclohexylammonium-salt/4580?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart