CH6371379886 Chem Impex International, Inc.

Fmoc-1,4-diaminobutane hydrochloride

  • MDL number MFCD01631652
  • Cas Number : 321660-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-NH(CH2)4NH2 HCl
Purity 99
Storage temp. Store at = -4
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-1-4-diaminobutane-hydrochloride/4583?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-1-4-diaminobutane-hydrochloride/4583?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart