CH6371379888 Chem Impex International, Inc.

Fmoc-1,6-diaminohexane hydrochloride

  • MDL number MFCD00830743
  • Cas Number : 166410-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 98 �dec.)
Synonym Fmoc-NH(CH2)6NH2 HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-1-6-diaminohexane-hydrochloride/4585?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-1-6-diaminohexane-hydrochloride/4585?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart