CH6371379889 Chem Impex International, Inc.

Trityl-1,2-ethylenediamine acetic acid salt

  • MDL number MFCD01631654
  • Cas Number : 1170595-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/trityl-1-2-diaminoethane-font-size-2-face-symbol-middot-font-acoh/4586?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/trityl-1-2-diaminoethane-font-size-2-face-symbol-middot-font-acoh/4586?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart