CH6371379890 Chem Impex International, Inc.

Trityl 1,3-diaminopropane acetate

  • MDL number MFCD01631655
  • Cas Number : 325143-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-143 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/trityl-1-3-diaminopropane-acetate/4587?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/trityl-1-3-diaminopropane-acetate/4587?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart