CH6371379891 Chem Impex International, Inc.

Trityl-1,4-diaminobutane acetate

  • MDL number MFCD01631653

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-125
Synonym Trt-NH(CH2)4NH2 AcOH
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/trityl-1-4-diaminobutane-acetate/4588?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/trityl-1-4-diaminobutane-acetate/4588?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart