CH6371379892 Chem Impex International, Inc.

Trityl-1,6-diaminohexane�H

  • MDL number MFCD01632434
  • Cas Number : 325708-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-127
Synonym Trt-NH(CH2)6NH2 AcOH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/trityl-1-6-diaminohexane-font-size-2-face-symbol-middot-font-acoh/4589?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/trityl-1-6-diaminohexane-font-size-2-face-symbol-middot-font-acoh/4589?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart