CH6371379893 Chem Impex International, Inc.

Boc-1,5-diaminopentane p-toluenesulfonate

  • MDL number MFCD01631657
  • Cas Number : 713520-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-137
Synonym Boc-NH(CH2)5NH2 TosOH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-1-5-diaminopentane-p-toluenesulfonate/4590?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-1-5-diaminopentane-p-toluenesulfonate/4590?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart