CH6371379895 Chem Impex International, Inc.

Fmoc-7-aminoheptanoic acid

  • MDL number MFCD01861353
  • Cas Number : 127582-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-157�C
Synonym Fmoc-7-Ahp-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-7-aminoheptanoic-acid/4592?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-7-aminoheptanoic-acid/4592?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart