CH6371379904 Chem Impex International, Inc.

L-Cyclopentylglycine

  • MDL number MFCD03788076
  • Cas Number : 2521-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Gly(cyclopentyl)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-cyclopentylglycine/4602?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-cyclopentylglycine/4602?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart