CH6371379923 Chem Impex International, Inc.

Boc-L-asparaginol

  • MDL number MFCD00270249
  • Cas Number : 30044-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Nalpha-Boc-L-Asn-ol
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-asparaginol/4629?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-asparaginol/4629?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart