CH6371379934 Chem Impex International, Inc.

Fmoc-D-cyclopentylglycine

  • MDL number MFCD01311778
  • Cas Number : 136555-16-3
  • Molecular Weight : 365.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 190 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Crystals or white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-cyclopentylglycine/4647?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-cyclopentylglycine/4647?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart